Assay Details
| Gene Symbol: Stac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016853.2 | NP_058549.1 | 5 - 6 | 792 | 64 | |
| GenBank mRNA | AK134777.1 | - | 4 - 5 | 522 | 64 | |
| AK143440.1 | - | 5 - 6 | 825 | 64 | ||
| BC107325.1 | - | 5 - 6 | 716 | 40054255 | 64 | |
| BC107326.1 | - | 5 - 6 | 716 | 40054256 | 64 | |
| D86639.1 | - | 5 - 6 | 792 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stac
Genomic Map