Assay Details
| Gene Symbol: Porcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036746.1 | - | 8 - 9 | 1007 | 69 | |
| AB036747.1 | - | 7 - 8 | 989 | 69 | ||
| AB036748.1 | - | 8 - 9 | 1004 | 69 | ||
| AB036749.1 | - | 9 - 10 | 1022 | 69 | ||
| AK010405.1 | - | 4 - 5 | 461 | 69 | ||
| BC027292.1 | - | 7 - 8 | 870 | 69 | ||
| BC032284.1 | - | 8 - 9 | 974 | 5362485 | 69 | |
| BU707440.1 | - | 5 - 6 | 439 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Porcn
Genomic Map