Assay Details
| Gene Symbol: Porcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036746.1 | - | 10 - 11 | 1177 | 63 | |
| AB036747.1 | - | 9 - 10 | 1159 | 63 | ||
| AB036748.1 | - | 10 - 11 | 1174 | 63 | ||
| AB036749.1 | - | 11 - 12 | 1192 | 63 | ||
| AK010405.1 | - | 6 - 7 | 631 | 63 | ||
| BC027292.1 | - | 9 - 10 | 1040 | 63 | ||
| BC032284.1 | - | 10 - 11 | 1144 | 5362485 | 63 | |
| BU707440.1 | - | 7 - 8 | 609 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Porcn
Genomic Map