Assay Details
| Gene Symbol: Porcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036746.1 | - | 11 - 12 | 1246 | 69 | |
| AB036747.1 | - | 10 - 11 | 1228 | 69 | ||
| AB036748.1 | - | 11 - 12 | 1243 | 69 | ||
| AB036749.1 | - | 12 - 13 | 1261 | 69 | ||
| AK010405.1 | - | 7 - 8 | 700 | 69 | ||
| BC027292.1 | - | 10 - 11 | 1109 | 69 | ||
| BC032284.1 | - | 11 - 12 | 1213 | 5362485 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Porcn
Genomic Map