Assay Details
| Gene Symbol: Prg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016914.2 | NP_058610.1 | 1 - 2 | 91 | 70 | |
| GenBank mRNA | AF202533.1 | - | 1 - 2 | 91 | 70 | |
| AK158110.1 | - | 1 - 2 | 111 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF202533.1,AK158110.1,NM_016914.2 transcripts.
Set Membership: |
|

Prg3
Genomic Map