Assay Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016985.2 | NP_058681.1 | 9 - 10 | 1225 | 77 | |
GenBank mRNA | AF073997.1 | - | 9 - 10 | 1198 | 77 | |
AK051390.1 | - | 2 - 3 | 223 | 77 | ||
BC056376.1 | - | 9 - 10 | 1225 | 6849757 | 77 | |
BC057337.1 | - | 9 - 10 | 1219 | 6825916 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
