Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 1 - 2 | 36 | 56 | |
| GenBank mRNA | BB610735.1 | - | 1 - 2 | 36 | 56 | |
| BC140998.1 | - | 1 - 2 | 37 | 9055610 | 56 | |
| BC145081.1 | - | 1 - 2 | 37 | 9053629 | 56 | |
| M37030.1 | - | 1 - 2 | 43 | 56 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC140998.1,BC145081.1,M37030.1,NM_017461.2 transcripts.
Set Membership: |
|

Sept1
Genomic Map