Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017461.2 | NP_059489.2 | 9 - 10 | 962 | 88 | |
| GenBank mRNA | AV507599.1 | - | 1 - 2 | 95 | 88 | |
| AY974598.1 | - | 1 - 2 | 82 | 88 | ||
| BC140998.1 | - | 9 - 10 | 963 | 9055610 | 88 | |
| BC145081.1 | - | 9 - 10 | 963 | 9053629 | 88 | |
| BX523183.1 | - | 2 - 3 | 278 | 88 | ||
| M37030.1 | - | 9 - 10 | 966 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept1
Genomic Map