Assay Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017462.2 | NP_059490.2 | 13 - 14 | 2567 | 75 | |
| GenBank mRNA | AB121698.1 | - | 12 - 13 | 2260 | 75 | |
| AK133584.1 | - | 13 - 14 | 2493 | 75 | ||
| BC053024.1 | - | 13 - 14 | 2493 | 75 | ||
| BC080291.1 | - | 13 - 14 | 2398 | 75 | ||
| U53584.1 | - | 13 - 14 | 2487 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polg
Genomic Map