Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 4 - 5 | 450 | 95 | |
| GenBank mRNA | AB028921.1 | - | 4 - 5 | 443 | 95 | |
| AK011325.1 | - | 4 - 5 | 455 | 95 | ||
| AK166526.1 | - | 5 - 6 | 366 | 95 | ||
| BC046801.1 | - | 4 - 5 | 439 | 6403966 | 95 | |
| BC051015.1 | - | 4 - 5 | 450 | 95 | ||
More Information
Set Membership: |
|

Akap8l
Genomic Map