Assay Details
| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018733.2 | NP_061203.2 | 21 - 22 | 4159 | 94 | |
| GenBank mRNA | L42339.1 | - | 5 - 6 | 912 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scn1a
Genomic Map