Assay Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018750.3 | NP_061220.2 | 4 - 5 | 1020 | 88 | |
| GenBank mRNA | AF053959.1 | - | 4 - 5 | 1002 | 88 | |
| AK005059.2 | - | 4 - 5 | 1016 | 88 | ||
| AK088751.1 | - | 3 - 4 | 655 | 88 | ||
| AK131147.1 | - | 3 - 4 | 613 | 88 | ||
| AK155534.1 | - | 4 - 5 | 1016 | 88 | ||
| AK155869.1 | - | 3 - 4 | 692 | 88 | ||
| AY261333.1 | - | 3 - 4 | 631 | 88 | ||
| BC089605.1 | - | 4 - 5 | 1022 | 30121804 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rassf5
Genomic Map