Assay Details
| Gene Symbol: Cpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018755.2 | NP_061225.2 | 4 - 5 | 937 | 76 | |
| NM_176073.4 | NP_788262.1 | 3 - 4 | 847 | 76 | ||
| GenBank mRNA | AF009513.1 | - | 3 - 4 | 728 | 76 | |
| AF107835.1 | - | 3 - 4 | 815 | 76 | ||
| AK032972.1 | - | 3 - 4 | 712 | 76 | ||
| AK075686.1 | - | 3 - 4 | 778 | 76 | ||
| AK135096.1 | - | 3 - 4 | 834 | 76 | ||
| AK137164.1 | - | 3 - 4 | 721 | 76 | ||
| AK152145.1 | - | 4 - 5 | 815 | 76 | ||
| AK154361.1 | - | 3 - 4 | 457 | 76 | ||
| BC010977.1 | - | 3 - 4 | 832 | 76 | ||
| BC017373.1 | - | 3 - 4 | 850 | 76 | ||
| BC037067.1 | - | 3 - 4 | 694 | 4989536 | 76 | |
More Information
Set Membership: |
|

Cpq
Genomic Map