Assay Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018775.4 | NP_061245.3 | 9 - 10 | 1739 | 65 | |
| GenBank mRNA | AK138630.1 | - | 9 - 10 | 1737 | 65 | |
| AK141520.1 | - | 9 - 10 | 1728 | 65 | ||
| AK141816.1 | - | 9 - 10 | 1693 | 65 | ||
| AK162371.1 | - | 3 - 4 | 514 | 65 | ||
| BC005421.1 | - | 6 - 7 | 1074 | 65 | ||
| BC065081.1 | - | 9 - 10 | 1673 | 6832495 | 65 | |
| U88873.1 | - | 5 - 6 | 959 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d8
Genomic Map