Assay Details
| Gene Symbol: St3gal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018784.2 | NP_061254.1 | 1 - 2 | 237 | 115 | |
| GenBank mRNA | AF119390.1 | - | 1 - 2 | 238 | 115 | |
| AK041173.1 | - | 1 - 2 | 280 | 115 | ||
| AK048950.1 | - | 1 - 2 | 281 | 115 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF119390.1,AK041173.1,NM_018784.2 transcripts.
Set Membership: |
|

St3gal6
Genomic Map