Assay Details
| Gene Symbol: St3gal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018784.2 | NP_061254.1 | 4 - 5 | 662 | 105 | |
| GenBank mRNA | AB063326.1 | - | 2 - 3 | 165 | 105 | |
| AF119390.1 | - | 4 - 5 | 662 | 105 | ||
| AK028442.1 | - | 4 - 5 | 481 | 105 | ||
| AK033562.1 | - | 4 - 5 | 549 | 105 | ||
| AK041173.1 | - | 4 - 5 | 705 | 105 | ||
| AK048950.1 | - | 4 - 5 | 706 | 105 | ||
| AK162619.1 | - | 5 - 6 | 642 | 105 | ||
| AK177361.1 | - | 3 - 4 | 409 | 105 | ||
| BC052338.1 | - | 3 - 4 | 423 | 30061291 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

St3gal6
Genomic Map