Assay Details
| Gene Symbol: St3gal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018784.2 | NP_061254.1 | 5 - 6 | 774 | 102 | |
| GenBank mRNA | AF119390.1 | - | 5 - 6 | 774 | 102 | |
| AK028442.1 | - | 5 - 6 | 593 | 102 | ||
| AK033562.1 | - | 5 - 6 | 661 | 102 | ||
| AK041173.1 | - | 5 - 6 | 817 | 102 | ||
| AK048950.1 | - | 5 - 6 | 818 | 102 | ||
| AK162619.1 | - | 6 - 7 | 754 | 102 | ||
| AK197012.1 | - | 1 - 2 | 50 | 102 | ||
| AK207906.1 | - | 1 - 2 | 50 | 102 | ||
| BC052338.1 | - | 4 - 5 | 535 | 30061291 | 102 | |
More Information
Set Membership: |
|

St3gal6
Genomic Map