Assay Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018857.1 | NP_061345.1 | 1 - 2 | 198 | 83 | |
| GenBank mRNA | AK144391.1 | - | 2 - 3 | 130 | 83 | |
| AK146799.1 | - | 1 - 2 | 248 | 83 | ||
| BC023753.1 | - | 1 - 2 | 202 | 5354484 | 83 | |
| D86370.1 | - | 1 - 2 | 198 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msln
Genomic Map