Assay Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018857.1 | NP_061345.1 | 5 - 6 | 495 | 61 | |
| GenBank mRNA | AK144391.1 | - | 6 - 7 | 427 | 61 | |
| AK146799.1 | - | 5 - 6 | 545 | 61 | ||
| BC023753.1 | - | 5 - 6 | 499 | 5354484 | 61 | |
| D86370.1 | - | 5 - 6 | 495 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | D86370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | D86370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762391 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146977 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | D86370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | D86370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31106053 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146977 | - | - |
| Whole Mouse Genome G4122A | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | D86370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | BC023753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | D86370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | NM_018857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | AK144391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146560 | AK146799.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126105 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591785 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591782 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13591802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31105991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31106049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13530527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13591804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13437857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13546601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13591793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31105995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13429707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31105991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31105993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31105998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13591814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13546615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13591806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13437859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13591809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13369123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31106037 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449012 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460238_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449000 | - | - |
| Mouse Genome 430 2.0 Array | 1460238_at | - | - |
| Mouse Genome 430 2.0 Array | 1460238_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56047_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56047_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700015250.mm.1 | NM_018857.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015250.mm.1 | NM_018857.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700015250.mm.1 | TC1700001671.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460238_PM_at | - | - |
| Mouse Expression Set 430 | 1460238_at | - | - |
| Murine Genome U74v2 Set | 102953_at | - | - |
| Murine Genome U74v2 Set | 102953_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460238_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460238_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460238_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460238_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 661148 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2640048 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231379 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231379 | - | - |
| mouse-6 v1.1 (Illumina) | scl50176.16.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50176.16.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231379 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231379 | - | - |

Msln
Genomic Map