Assay Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019482.2 | NP_062355.2 | 3 - 4 | 986 | 70 | |
| GenBank mRNA | AF207817.1 | - | 3 - 4 | 584 | 70 | |
| AK014033.1 | - | 3 - 4 | 969 | 70 | ||
| AK053871.1 | - | 3 - 4 | 971 | 70 | ||
| AK089764.1 | - | 3 - 4 | 932 | 70 | ||
| AK156194.1 | - | 3 - 4 | 971 | 70 | ||
| AK212589.1 | - | 1 - 2 | 232 | 70 | ||
| BC020413.1 | - | 3 - 4 | 917 | 70 | ||
| BC049074.1 | - | 3 - 4 | 986 | 6809247 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Panx1
Genomic Map