Assay Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019501.3 | NP_062374.2 | 5 - 6 | 706 | 99 | |
| GenBank mRNA | AB210841.1 | - | 5 - 6 | 626 | 99 | |
| AF118855.1 | - | 3 - 4 | 401 | 99 | ||
| AK012307.1 | - | 5 - 6 | 634 | 99 | ||
| BC026820.1 | - | 5 - 6 | 608 | 4039464 | 99 | |
| BC107273.1 | - | 5 - 6 | 521 | 99 | ||
| BC107274.1 | - | 5 - 6 | 525 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdss1
Genomic Map