Assay Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019501.3 | NP_062374.2 | 6 - 7 | 811 | 119 | |
| GenBank mRNA | AB210841.1 | - | 6 - 7 | 731 | 119 | |
| AF118855.1 | - | 4 - 5 | 506 | 119 | ||
| AK012307.1 | - | 6 - 7 | 739 | 119 | ||
| BC026820.1 | - | 6 - 7 | 713 | 4039464 | 119 | |
| BC107273.1 | - | 6 - 7 | 626 | 119 | ||
| BC107274.1 | - | 6 - 7 | 630 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdss1
Genomic Map