Assay Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019501.3 | NP_062374.2 | 8 - 9 | 1002 | 96 | |
| GenBank mRNA | AB210841.1 | - | 8 - 9 | 922 | 96 | |
| AF118855.1 | - | 6 - 7 | 697 | 96 | ||
| BC026820.1 | - | 8 - 9 | 904 | 4039464 | 96 | |
| BC107273.1 | - | 8 - 9 | 817 | 96 | ||
| BC107274.1 | - | 8 - 9 | 821 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdss1
Genomic Map