Assay Details
| Gene Symbol: Pitpnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301643.1 | NP_001288572.1 | 7 - 8 | 575 | 72 | |
| NM_001301644.1 | NP_001288573.1 | 7 - 8 | 544 | 72 | ||
| NM_019640.5 | NP_062614.1 | 7 - 8 | 575 | 72 | ||
| GenBank mRNA | AK028235.1 | - | 7 - 8 | 510 | 72 | |
| BC034676.1 | - | 7 - 8 | 465 | 4481685 | 72 | |
| CK619028.1 | - | 7 - 8 | 458 | 72 | ||
| U46934.1 | - | 7 - 8 | 477 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnb
Genomic Map