Assay Details
| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019642.4 | NP_062616.2 | 12 - 13 | 1719 | 100 | |
| GenBank mRNA | AK004968.1 | - | 12 - 13 | 1720 | 100 | |
| AK018761.2 | - | 12 - 13 | 1759 | 100 | ||
| AK153952.1 | - | 12 - 13 | 1580 | 100 | ||
| AK161624.1 | - | 12 - 13 | 1530 | 100 | ||
| AK166778.1 | - | 12 - 13 | 1505 | 100 | ||
| AK167864.1 | - | 12 - 13 | 1499 | 100 | ||
| AK167866.1 | - | 12 - 13 | 1499 | 100 | ||
| AK168221.1 | - | 12 - 13 | 1499 | 100 | ||
| BC010509.1 | - | 11 - 12 | 1492 | 100 | ||
| BC046806.1 | - | 12 - 13 | 1747 | 6829209 | 100 | |
| D31717.1 | - | 11 - 12 | 1505 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn2
Genomic Map