Assay Details
| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252580.1 | NP_001239509.1 | 9 - 10 | 1252 | 69 | |
| NM_019729.3 | NP_062703.2 | 9 - 10 | 1219 | 69 | ||
| GenBank mRNA | AB045709.1 | - | 8 - 9 | 993 | 69 | |
| AF057146.1 | - | 8 - 9 | 1010 | 69 | ||
| AK122195.1 | - | 9 - 10 | 1130 | 69 | ||
| AK129003.1 | - | 9 - 10 | 1220 | 69 | ||
| AK134058.1 | - | 9 - 10 | 1174 | 69 | ||
| AK142881.1 | - | 6 - 7 | 717 | 69 | ||
| AK153015.1 | - | 8 - 9 | 967 | 69 | ||
| AK163469.1 | - | 9 - 10 | 1152 | 69 | ||
| BC038123.1 | - | 9 - 10 | 1204 | 69 | ||
| BC050947.1 | - | 9 - 10 | 1184 | 6825981 | 69 | |
| BC061465.1 | - | 9 - 10 | 1184 | 6825981 | 69 | |
| BC066126.1 | - | 9 - 10 | 1182 | 30301665 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp8
Genomic Map