Assay Details
| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252580.1 | NP_001239509.1 | 12 - 13 | 2118 | 89 | |
| NM_019729.3 | NP_062703.2 | 12 - 13 | 2085 | 89 | ||
| GenBank mRNA | AB045709.1 | - | 11 - 12 | 1859 | 89 | |
| AF057146.1 | - | 11 - 12 | 1876 | 89 | ||
| AK122195.1 | - | 12 - 13 | 1996 | 89 | ||
| AK129003.1 | - | 12 - 13 | 2086 | 89 | ||
| AK134058.1 | - | 12 - 13 | 2040 | 89 | ||
| AK142881.1 | - | 9 - 10 | 1583 | 89 | ||
| AK153015.1 | - | 11 - 12 | 1833 | 89 | ||
| AK166540.1 | - | 10 - 11 | 1439 | 89 | ||
| BC027052.1 | - | 2 - 3 | 638 | 89 | ||
| BC050947.1 | - | 12 - 13 | 2050 | 6825981 | 89 | |
| BC061465.1 | - | 12 - 13 | 2050 | 6825981 | 89 | |
| BC066126.1 | - | 12 - 13 | 2048 | 30301665 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp8
Genomic Map