Assay Details
| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033988.2 | NP_001029160.1 | 2 - 3 | 281 | 69 | |
| NM_001284319.1 | NP_001271248.1 | 2 - 3 | 268 | 69 | ||
| NM_019744.4 | NP_062718.2 | 2 - 3 | 191 | 69 | ||
| GenBank mRNA | AF159461.1 | - | 2 - 3 | 188 | 69 | |
| AK014479.1 | - | 2 - 3 | 189 | 69 | ||
| AK129020.1 | - | 2 - 3 | 239 | 69 | ||
| AK132981.1 | - | 69 | ||||
| AK143965.1 | - | 2 - 3 | 186 | 69 | ||
| AK145070.1 | - | 2 - 3 | 242 | 69 | ||
| AK145945.1 | - | 2 - 3 | 234 | 69 | ||
| AK145954.1 | - | 2 - 3 | 238 | 69 | ||
| AK146384.1 | - | 2 - 3 | 192 | 69 | ||
| AK146498.1 | - | 2 - 3 | 231 | 69 | ||
| AK146598.1 | - | 2 - 3 | 215 | 69 | ||
| AK146646.1 | - | 2 - 3 | 186 | 69 | ||
| AK146649.1 | - | 2 - 3 | 188 | 69 | ||
| AK147036.1 | - | 2 - 3 | 189 | 69 | ||
| AK147125.1 | - | 2 - 3 | 233 | 69 | ||
| AK156999.1 | - | 2 - 3 | 267 | 69 | ||
| BC031528.1 | - | 2 - 3 | 232 | 4008986 | 69 | |
| BC085086.1 | - | 2 - 3 | 177 | 6335192 | 69 | |
| CF161810.1 | - | 2 - 3 | 276 | 69 | ||
| CJ088075.1 | - | 2 - 3 | 284 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001284319.1 transcripts.
Set Membership: |
|

Ncoa4
Genomic Map