Assay Details
| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033988.2 | NP_001029160.1 | 8 - 9 | 1871 | 87 | |
| NM_001284319.1 | NP_001271248.1 | 8 - 9 | 1863 | 87 | ||
| NM_019744.4 | NP_062718.2 | 8 - 9 | 1781 | 87 | ||
| GenBank mRNA | AF159461.1 | - | 8 - 9 | 1778 | 87 | |
| AK014479.1 | - | 8 - 9 | 1782 | 87 | ||
| AK016355.1 | - | 1 - 2 | 49 | 87 | ||
| AK032951.1 | - | 6 - 7 | 1537 | 87 | ||
| AK129020.1 | - | 8 - 9 | 1829 | 87 | ||
| AK143965.1 | - | 8 - 9 | 1775 | 87 | ||
| AK145945.1 | - | 8 - 9 | 1824 | 87 | ||
| AK145954.1 | - | 8 - 9 | 1828 | 87 | ||
| AK146384.1 | - | 8 - 9 | 1782 | 87 | ||
| AK146498.1 | - | 8 - 9 | 1821 | 87 | ||
| AK146598.1 | - | 8 - 9 | 1805 | 87 | ||
| AK146646.1 | - | 8 - 9 | 1776 | 87 | ||
| AK146649.1 | - | 8 - 9 | 1778 | 87 | ||
| AK147036.1 | - | 8 - 9 | 1780 | 87 | ||
| AK147125.1 | - | 8 - 9 | 1823 | 87 | ||
| AK156999.1 | - | 8 - 9 | 1863 | 87 | ||
| BC085086.1 | - | 8 - 9 | 1767 | 6335192 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa4
Genomic Map