Assay Details
| Gene Symbol: Vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019780.1 | NP_062754.1 | 1 - 2 | 65 | 74 | |
| GenBank mRNA | AB578904.1 | - | 1 - 2 | 80 | 74 | |
| AB578905.1 | - | 1 - 2 | 69 | 74 | ||
| AB578906.1 | - | 1 - 2 | 63 | 74 | ||
| AB578907.1 | - | 1 - 2 | 62 | 74 | ||
| AB578909.1 | - | 1 - 2 | 79 | 74 | ||
| AB578910.1 | - | 1 - 2 | 48 | 74 | ||
| AB578911.1 | - | 1 - 2 | 68 | 74 | ||
| AF193794.1 | - | 1 - 2 | 65 | 74 | ||
| AK036426.1 | - | 1 - 2 | 53 | 74 | ||
| AK150330.1 | - | 1 - 2 | 75 | 74 | ||
| BC005663.1 | - | 1 - 2 | 54 | 2645432 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF193794.1,AK150330.1,BC005663.1,NM_019780.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049902 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049907 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044202 | AK150330.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044202 | BC005663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044202 | AB578901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044202 | AF193794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044202 | AK008248.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044197 | AK150330.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044197 | BC005663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044197 | AB578901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044197 | AF193794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044197 | AK008248.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495530 | AF373410.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495530 | NM_130903.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50311 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168846 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044202 | AK150330.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044202 | BC005663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044202 | AB578901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044202 | AF193794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044202 | AK008248.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044197 | AK150330.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044197 | BC005663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044197 | AB578901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044197 | AF193794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044197 | AK008248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264554 | AK150330.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264554 | AB578901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264554 | AF193794.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264554 | AK008248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264554 | AK036426.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049920 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05724378 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168846 | - | - |
| Whole Mouse Genome G4122A | A_52_P83688 | AK044468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456071 | AK150330.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456071 | AB578901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456071 | AF193794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456071 | AK004103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456071 | AB578899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P649864 | AK036426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264554 | AK150330.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264554 | AB578901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264554 | AF193794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264554 | AK008248.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264554 | AK036426.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100891 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168846 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04782189 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04782193 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049907 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04550926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04535359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04243007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04535358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04415574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525382 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440641_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455302_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417660_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417659_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525381 | - | - |
| Mouse Genome 430 2.0 Array | 1417659_at | - | - |
| Mouse Genome 430 2.0 Array | 1455302_at | - | - |
| Mouse Genome 430 2.0 Array | 1440641_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440641_at | - | - |
| Mouse Genome 430 2.0 Array | 1417660_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455302_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417660_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417659_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56433_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56433_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500012270.mm.1 | TC0500001389.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500012270.mm.1 | NM_019780.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500012270.mm.1 | NM_019780.1 | Refseq |
| Mouse Expression Set 430 | 1455302_at | - | - |
| Mouse Expression Set 430 | 1417659_at | - | - |
| Mouse Expression Set 430 | 1455302_PM_at | - | - |
| Mouse Expression Set 430 | 1417659_PM_at | - | - |
| Mouse Expression Set 430 | 1417660_PM_s_at | - | - |
| Mouse Expression Set 430 | 1417660_s_at | - | - |
| Mouse Expression Set 430 | 1440641_PM_at | - | - |
| Mouse Expression Set 430 | 1440641_at | - | - |
| Murine Genome U74v2 Set | 97876_at | - | - |
| Murine Genome U74v2 Set | 97876_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455302_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417660_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455302_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417659_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417660_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417659_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417659_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455302_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417660_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417660_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455302_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417659_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440641_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440641_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 924699 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260497 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2495644 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2446175 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2495644 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2446175 | - | - |
| mouse-6 v1.1 (Illumina) | scl056433.4_275-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0004181.1_42-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580687 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004181.1_42-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056433.4_275-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260497 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2495644 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2446175 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2955883 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2495644 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2955883 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2446175 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056433.4_275-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004181.1_42-S | - | - |

Vps29
Genomic Map