Assay Details
| Gene Symbol: Tmeff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019790.4 | NP_062764.1 | 6 - 7 | 1111 | 103 | |
| GenBank mRNA | AB017270.1 | - | 6 - 7 | 1074 | 103 | |
| AB041565.1 | - | 6 - 7 | 1116 | 103 | ||
| AK029307.1 | - | 6 - 7 | 1111 | 103 | ||
| AK076254.1 | - | 6 - 7 | 1111 | 103 | ||
| AK135038.1 | - | 6 - 7 | 1098 | 103 | ||
| BC034850.1 | - | 6 - 7 | 1087 | 1397175 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmeff2
Genomic Map