Assay Details
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019810.4 | NP_062784.3 | 4 - 5 | 612 | 71 | |
| GenBank mRNA | AF163846.1 | - | 4 - 5 | 374 | 71 | |
| AF208031.1 | - | 4 - 5 | 612 | 71 | ||
| AK033512.1 | - | 4 - 5 | 598 | 71 | ||
| AK085602.1 | - | 4 - 5 | 403 | 71 | ||
| BC003845.1 | - | 4 - 5 | 592 | 3497611 | 71 | |
| U27269.1 | - | 2 - 3 | 182 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a1
Genomic Map