Assay Details
| Gene Symbol: Trpc4ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163452.1 | NP_001156924.1 | 9 - 10 | 1294 | 71 | |
| NM_019828.2 | NP_062802.2 | 9 - 10 | 1270 | 71 | ||
| GenBank mRNA | AF093589.1 | - | 5 - 6 | 766 | 71 | |
| AF130458.1 | - | 9 - 10 | 1218 | 71 | ||
| AF509881.1 | - | 9 - 10 | 1216 | 71 | ||
| AK140799.1 | - | 8 - 9 | 982 | 71 | ||
| AK154263.1 | - | 9 - 10 | 71 | |||
| AK156047.1 | - | 9 - 10 | 1243 | 71 | ||
| AK165259.1 | - | 9 - 10 | 1243 | 71 | ||
| AK170085.1 | - | 9 - 10 | 1232 | 71 | ||
| AK171404.1 | - | 9 - 10 | 1243 | 71 | ||
| AK220239.1 | - | 9 - 10 | 1270 | 71 | ||
| BC003931.1 | - | 2 - 3 | 230 | 71 | ||
| BC033274.1 | - | 9 - 10 | 1222 | 3673725 | 71 | |
| BC057330.1 | - | 9 - 10 | 1252 | 6816155 | 71 | |
More Information
Set Membership: |
|

Trpc4ap
Genomic Map