Assay Details
| Gene Symbol: Trpc4ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163452.1 | NP_001156924.1 | 11 - 12 | 1485 | 89 | |
| NM_019828.2 | NP_062802.2 | 11 - 12 | 1461 | 89 | ||
| GenBank mRNA | AF093589.1 | - | 7 - 8 | 957 | 89 | |
| AF130458.1 | - | 11 - 12 | 1409 | 89 | ||
| AF509881.1 | - | 11 - 12 | 1407 | 89 | ||
| AK140799.1 | - | 10 - 11 | 1191 | 89 | ||
| AK154263.1 | - | 11 - 12 | 1434 | 89 | ||
| AK156047.1 | - | 11 - 12 | 1434 | 89 | ||
| AK165259.1 | - | 11 - 12 | 1434 | 89 | ||
| AK170085.1 | - | 11 - 12 | 1423 | 89 | ||
| AK171404.1 | - | 11 - 12 | 1434 | 89 | ||
| AK182119.1 | - | 1 - 2 | 59 | 89 | ||
| AK190064.1 | - | 1 - 2 | 59 | 89 | ||
| AK192661.1 | - | 1 - 2 | 58 | 88 | ||
| AK195086.1 | - | 1 - 2 | 59 | 89 | ||
| AK220239.1 | - | 11 - 12 | 1461 | 89 | ||
| BC003931.1 | - | 4 - 5 | 421 | 89 | ||
| BC033274.1 | - | 11 - 12 | 1413 | 3673725 | 89 | |
| BC057330.1 | - | 11 - 12 | 1443 | 6816155 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpc4ap
Genomic Map