Assay Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019879.3 | NP_063932.2 | 3 - 4 | 476 | 70 | |
| GenBank mRNA | AF144101.2 | - | 3 - 4 | 309 | 70 | |
| AK002754.1 | - | 3 - 4 | 311 | 70 | ||
| AK005080.1 | - | 3 - 4 | 325 | 70 | ||
| AK184184.1 | - | 3 - 4 | 252 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Suclg1
Genomic Map