Assay Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019879.3 | NP_063932.2 | 5 - 6 | 737 | 85 | |
| GenBank mRNA | AF144101.2 | - | 5 - 6 | 570 | 85 | |
| AK005080.1 | - | 5 - 6 | 586 | 85 | ||
| AK088888.1 | - | 5 - 6 | 594 | 85 | ||
| BC011087.1 | - | 5 - 6 | 558 | 4012310 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Suclg1
Genomic Map