Assay Details
| Gene Symbol: Ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019930.2 | NP_064314.2 | 11 - 12 | 1786 | 65 | |
| GenBank mRNA | AF006465.1 | - | 12 - 13 | 1615 | 65 | |
| AK132714.1 | - | 11 - 12 | 1265 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp9
Genomic Map