Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 4 - 5 | 282 | 105 | |
| GenBank mRNA | AF139179.1 | - | 4 - 5 | 286 | 105 | |
| AK050619.1 | - | 5 - 6 | 309 | 105 | ||
| AK083379.1 | - | 5 - 6 | 309 | 105 | ||
| AK188434.1 | - | 2 - 3 | 89 | 106 | ||
| AK212629.1 | - | 3 - 4 | 139 | 105 | ||
| BC052702.1 | - | 4 - 5 | 274 | 6400922 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map