Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 6 - 7 | 474 | 83 | |
| GenBank mRNA | AF139179.1 | - | 6 - 7 | 478 | 83 | |
| AK050619.1 | - | 7 - 8 | 498 | 83 | ||
| AK083379.1 | - | 7 - 8 | 498 | 83 | ||
| AK210270.1 | - | 3 - 4 | 258 | 83 | ||
| BC052702.1 | - | 6 - 7 | 463 | 6400922 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map