Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 8 - 9 | 697 | 88 | |
| GenBank mRNA | AF139179.1 | - | 8 - 9 | 701 | 88 | |
| AK050619.1 | - | 9 - 10 | 721 | 88 | ||
| AK083379.1 | - | 9 - 10 | 721 | 88 | ||
| BC052702.1 | - | 8 - 9 | 686 | 6400922 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map