Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 13 - 14 | 1184 | 105 | |
| GenBank mRNA | AF139179.1 | - | 13 - 14 | 1188 | 105 | |
| AK050619.1 | - | 14 - 15 | 1208 | 105 | ||
| AK083379.1 | - | 14 - 15 | 1208 | 105 | ||
| AK207764.1 | - | 3 - 4 | 188 | 105 | ||
| BC052702.1 | - | 13 - 14 | 1173 | 6400922 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map