Assay Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020022.2 | NP_064406.1 | 4 - 5 | 415 | 78 | |
| GenBank mRNA | AK075997.1 | - | 4 - 5 | 327 | 78 | |
| AK154681.1 | - | 4 - 5 | 369 | 78 | ||
| BC004812.1 | - | 1 - 2 | 87 | 78 | ||
| BC023028.1 | - | 4 - 5 | 336 | 5374851 | 78 | |
| BC099370.1 | - | 4 - 5 | 379 | 30937690 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc2
Genomic Map