Assay Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020022.2 | NP_064406.1 | 7 - 8 | 778 | 71 | |
| GenBank mRNA | AK011649.1 | - | 7 - 8 | 692 | 71 | |
| AK075997.1 | - | 7 - 8 | 690 | 71 | ||
| AK154681.1 | - | 7 - 8 | 732 | 71 | ||
| BC004812.1 | - | 4 - 5 | 450 | 71 | ||
| BC023028.1 | - | 7 - 8 | 699 | 5374851 | 71 | |
| BC099370.1 | - | 7 - 8 | 742 | 30937690 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc2
Genomic Map