Assay Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020022.2 | NP_064406.1 | 8 - 9 | 844 | 98 | |
| GenBank mRNA | AK011353.1 | - | 8 - 9 | 760 | 98 | |
| AK011649.1 | - | 8 - 9 | 758 | 98 | ||
| AK075997.1 | - | 8 - 9 | 756 | 98 | ||
| AK154681.1 | - | 8 - 9 | 798 | 98 | ||
| BC004812.1 | - | 5 - 6 | 516 | 98 | ||
| BC023028.1 | - | 8 - 9 | 765 | 5374851 | 98 | |
| BC099370.1 | - | 8 - 9 | 808 | 30937690 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfc2
Genomic Map