Assay Details
| Gene Symbol: Rfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020022.2 | NP_064406.1 | 10 - 11 | 1032 | 70 | |
| GenBank mRNA | AK011353.1 | - | 10 - 11 | 948 | 70 | |
| AK011649.1 | - | 10 - 11 | 945 | 70 | ||
| AK075997.1 | - | 10 - 11 | 944 | 70 | ||
| AK154681.1 | - | 10 - 11 | 986 | 70 | ||
| BC004812.1 | - | 7 - 8 | 704 | 70 | ||
| BC023028.1 | - | 10 - 11 | 953 | 5374851 | 70 | |
| BC099370.1 | - | 10 - 11 | 996 | 30937690 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143823 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117164 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09573365 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09573356 | - | - |
| Whole Mouse Genome G4122A | A_52_P981680 | - | - |
| Whole Mouse Genome G4122A | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | AK154681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC004812.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC023028.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | BC099370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P254912 | NM_020022.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106708 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P33001057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P33001058 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526219 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457669_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457638_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417503_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526217 | - | - |
| Mouse Genome 430 2.0 Array | 1449606_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457638_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417503_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449606_at | - | - |
| Mouse Genome 430 2.0 Array | 1457638_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1457669_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1457669_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417503_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19718_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19718_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500013992.mm.1 | TC0500001560.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013992.mm.1 | NM_020022.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013992.mm.1 | NM_020022.2 | Refseq |
| Mouse Expression Set 430 | 1417503_at | - | - |
| Mouse Expression Set 430 | 1417503_PM_at | - | - |
| Mouse Expression Set 430 | 1457638_x_at | - | - |
| Mouse Expression Set 430 | 1457638_PM_x_at | - | - |
| Mouse Expression Set 430 | 1457669_x_at | - | - |
| Mouse Expression Set 430 | 1457669_PM_x_at | - | - |
| Murine Genome U74v2 Set | 160759_at | - | - |
| Murine Genome U74v2 Set | 160759_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417503_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417503_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417503_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417503_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457638_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457669_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457638_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457669_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 107040121 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106980288 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106980288 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2424690 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2749487 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734977 | - | - |
| mouse-6 v1.1 (Illumina) | scl27145.4.1_19-S | - | - |
| mouse-6 v1.1 (Illumina) | scl019718.11_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107040121 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106980288 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107040121 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106980288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2894497 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749487 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749484 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2894497 | - | - |

Rfc2
Genomic Map