Assay Details
| Gene Symbol: Poll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020032.2 | NP_064416.1 | 2 - 3 | 195 | 78 | |
| GenBank mRNA | AF176099.1 | - | 1 - 2 | 136 | 78 | |
| AJ131889.1 | - | 2 - 3 | 195 | 78 | ||
| AK133087.1 | - | 2 - 3 | 162 | 78 | ||
| BC004767.1 | - | 1 - 2 | 145 | 3156259 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poll
Genomic Map