Assay Details
| Gene Symbol: Slc15a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021301.3 | NP_067276.2 | 8 - 9 | 981 | 78 | |
| GenBank mRNA | AF111811.1 | - | 8 - 9 | 981 | 78 | |
| AK128939.1 | - | 8 - 9 | 814 | 78 | ||
| BC138627.1 | - | 8 - 9 | 938 | 8861647 | 78 | |
| BC138628.1 | - | 8 - 9 | 938 | 8861648 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a2
Genomic Map