Assay Details
| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025571.2 | NP_001020742.1 | 5 - 6 | 946 | 81 | |
| NM_021309.3 | NP_067284.1 | 5 - 6 | 946 | 81 | ||
| GenBank mRNA | AF203343.1 | - | 5 - 6 | 953 | 81 | |
| AK037489.1 | - | 5 - 6 | 668 | 81 | ||
| AK172182.1 | - | 5 - 6 | 698 | 81 | ||
| BC034847.1 | - | 5 - 6 | 678 | 3327113 | 81 | |
| U69490.1 | - | 5 - 6 | 656 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118141 | NM_001025571.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118141 | NM_021309.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118141 | AK037489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P118141 | BB154497.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163402 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128919 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118141 | NM_001025571.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118141 | NM_021309.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118141 | AK037489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P118141 | BB154497.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310332 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04979265 | - | - |
| Whole Mouse Genome G4122A | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226882 | NM_021309.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226882 | AF203343.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115713 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118141 | NM_001025571.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118141 | NM_021309.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118141 | AK037489.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118141 | BB154497.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02953054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02953055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02603317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02953057 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02603315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02603315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02659895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02356142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02967656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02513775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02392456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02606771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02606767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02953050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02953057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02953046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02953053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02709051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02603317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02709055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02606769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02709061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02606770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02884039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02646001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493077 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449105_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493076 | - | - |
| Mouse Genome 430 2.0 Array | 1449105_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449105_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27371_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27371_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300004737.mm.1 | NM_001025571.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004737.mm.1 | NM_001025571.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004737.mm.1 | NM_021309.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004737.mm.1 | NM_021309.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004737.mm.1 | TC0300000706.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449105_at | - | - |
| Mouse Expression Set 430 | 1449105_PM_at | - | - |
| Murine Genome U74v2 Set | 100292_at | - | - |
| Murine Genome U74v2 Set | 100292_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449105_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449105_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449105_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449105_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 783349 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105270181 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102230647 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2970594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2970594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102230647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228404 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2468455 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228404 | - | - |
| mouse-6 v1.1 (Illumina) | scl0027371.2_277-S | - | - |
| mouse-6 v1.1 (Illumina) | scl23017.5_177-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102230647 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0027371.2_277-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102230647 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228404 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228404 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0027371.2_277-S | - | - |

Sh2d2a
Genomic Map