| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
TC0700001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_001004468.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_001004468.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_021314.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_021314.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_206856.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016656.mm.1 |
NM_206856.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
TC0700001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_001004468.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_001004468.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_021314.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_021314.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_206856.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016654.mm.1 |
NM_206856.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
TC0700001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_001004468.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_001004468.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_021314.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_021314.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_206856.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016655.mm.1 |
NM_206856.3 |
Refseq |