Assay Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135727.2 | NP_001129199.1 | 3 - 4 | 702 | 122 | |
| NM_021389.6 | NP_067364.2 | 2 - 3 | 267 | 122 | ||
| NR_110969.1 | 2 - 3 | 267 | 122 | |||
| NR_110970.1 | 2 - 3 | 267 | 122 | |||
| GenBank mRNA | AK004636.1 | - | 2 - 3 | 267 | 122 | |
| AK018032.1 | - | 2 - 3 | 284 | 122 | ||
| AK020782.1 | - | 2 - 3 | 244 | 122 | ||
| AK030839.1 | - | 2 - 3 | 268 | 122 | ||
| AK038540.1 | - | 3 - 4 | 434 | 122 | ||
| AK049182.1 | - | 3 - 4 | 703 | 122 | ||
| AK080930.1 | - | 3 - 4 | 567 | 122 | ||
| AK172022.1 | - | 3 - 4 | 683 | 122 | ||
| BC034646.1 | - | 2 - 3 | 271 | 122 | ||
| BC052863.1 | - | 2 - 3 | 216 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3kbp1
Genomic Map